4-[(2-Bromo-4-chloro-3,5-dimethylphenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050355-500MG
050355-2.5G
101891-320EA
364.62
USD
101891-320
101891-322
4-[(2-Bromo-4-chloro-3,5-dimethylphenoxy)methyl]piperidine hydrochloride
4-[(2-Bromo-4-chloro-3,5-dimethylphenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₀BrCl₂NO |
MDL Number:
MFCD13560756 CAS Number: 1220032-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...