3-[(2-Bromo-4-chloro-3,5-dimethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050354-500MG 050354-2.5G
101891-316EA 364.62 USD
101891-316 101891-318
3-[(2-Bromo-4-chloro-3,5-dimethylphenoxy)methyl]piperidine hydrochloride
3-[(2-Bromo-4-chloro-3,5-dimethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀BrCl₂NO
MDL Number: MFCD13560755
CAS Number: 1220017-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR