4-(2-Bromo-4-chloro-3,5-dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050353-500MG 050353-2.5G
101891-312EA 364.62 USD
101891-312 101891-314
4-(2-Bromo-4-chloro-3,5-dimethylphenoxy)piperidine hydrochloride
4-(2-Bromo-4-chloro-3,5-dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈BrCl₂NO
MDL Number: MFCD13560754
CAS Number: 1220017-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR