3-(4-Benzyl-2-bromophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050351-500MG 050351-2.5G
101891-304EA 364.62 USD
101891-304 101891-306
3-(4-Benzyl-2-bromophenoxy)pyrrolidine hydrochloride
3-(4-Benzyl-2-bromophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉BrClNO
MDL Number: MFCD13560752
CAS Number: 1220017-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR