3-[(4-Benzyl-2-bromophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050349-500MG 050349-2.5G
101891-296EA 364.62 USD
101891-296 101891-298
3-[(4-Benzyl-2-bromophenoxy)methyl]piperidine hydrochloride
3-[(4-Benzyl-2-bromophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₃BrClNO
MDL Number: MFCD13560750
CAS Number: 1220035-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR