4-(4-Benzyl-2-bromophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050348-500MG 050348-2.5G
101891-292EA 364.62 USD
101891-292 101891-294
4-(4-Benzyl-2-bromophenoxy)piperidine hydrochloride
4-(4-Benzyl-2-bromophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁BrClNO
MDL Number: MFCD13560749
CAS Number: 1220032-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR