3-(4-Benzyl-2-bromophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050347-500MG 050347-2.5G
101891-288EA 364.62 USD
101891-288 101891-290
3-(4-Benzyl-2-bromophenoxy)piperidine hydrochloride
3-(4-Benzyl-2-bromophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁BrClNO
MDL Number: MFCD13560748
CAS Number: 1220017-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR