3-(4-Bromo-2-ethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050346-500MG 050346-2.5G
101891-284EA 364.62 USD
101891-284 101891-286
3-(4-Bromo-2-ethylphenoxy)pyrrolidine hydrochloride
3-(4-Bromo-2-ethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD13560747
CAS Number: 1220032-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR