3-[(4-Bromo-2-ethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050344-500MG 050344-2.5G
101891-276EA 364.62 USD
101891-276 101891-278
3-[(4-Bromo-2-ethylphenoxy)methyl]piperidine hydrochloride
3-[(4-Bromo-2-ethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁BrClNO
MDL Number: MFCD13560745
CAS Number: 1219967-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR