3-(4-Bromo-2-ethylphenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
050342-500MG
050342-2.5G
101891-268EA
364.62
USD
101891-268
101891-270
3-(4-Bromo-2-ethylphenoxy)piperidine hydrochloride
3-(4-Bromo-2-ethylphenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉BrClNO |
MDL Number:
MFCD13560743 CAS Number: 1220035-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...