2-Bromo-4-methoxyphenyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050340-500MG 050340-2.5G
101891-260EA 364.62 USD
101891-260 101891-262
2-Bromo-4-methoxyphenyl-4-piperidinylmethyl ether hydrochloride
2-Bromo-4-methoxyphenyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉BrClNO₂
MW: 336.66 g/mol
MDL Number: MFCD13560741
CAS Number: 1220017-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR