3-(2-Bromo-4-methoxyphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050337-500MG 050337-2.5G
101891-248EA 364.62 USD
101891-248 101891-250
3-(2-Bromo-4-methoxyphenoxy)piperidine hydrochloride
3-(2-Bromo-4-methoxyphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO₂
MDL Number: MFCD13560738
CAS Number: 1220017-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR