Methyl-3-[2-(3-piperidinyl)ethoxy]propanoate hydrochloride
Supplier: Matrix Scientific
050334-500MG
050334-2.5G
101891-236EA
364.62
USD
101891-236
101891-238
Methyl-3-[2-(3-piperidinyl)ethoxy]propanoate hydrochloride
Methyl-3-[2-(3-piperidinyl)ethoxy]propanoate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₂ClNO₃ |
MDL Number:
MFCD13560735 CAS Number: 1220032-59-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...