Cyclobutylmethyl-3-pyrrolidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050332-500MG 050332-2.5G
101891-228EA 364.62 USD
101891-228 101891-230
Cyclobutylmethyl-3-pyrrolidinylmethyl ether hydrochloride
Cyclobutylmethyl-3-pyrrolidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD13560733
CAS Number: 1220032-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR