3-[2-(Cyclobutylmethoxy)ethyl]piperidine hydrochloride
Supplier: Matrix Scientific
050330-500MG
050330-2.5G
101891-220EA
364.62
USD
101891-220
101891-222
3-[2-(Cyclobutylmethoxy)ethyl]piperidine hydrochloride
3-[2-(Cyclobutylmethoxy)ethyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₄ClNO |
MDL Number:
MFCD13560731 CAS Number: 1220016-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...