4-[2-(Cyclobutylmethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050329-500MG 050329-2.5G
101891-216EA 364.62 USD
101891-216 101891-218
4-[2-(Cyclobutylmethoxy)ethyl]piperidine hydrochloride
4-[2-(Cyclobutylmethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₄ClNO
MDL Number: MFCD13560730
CAS Number: 1220032-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR