2-(2-Isopropoxyethyl)piperidine hydrochloride

Supplier: Matrix Scientific

050327-500MG 050327-2.5G
101891-208EA 364.62 USD
101891-208 101891-210
2-(2-Isopropoxyethyl)piperidine hydrochloride
2-(2-Isopropoxyethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂ClNO
MW: 207.75 g/mol
MDL Number: MFCD13560728
CAS Number: 1219972-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR