4-(2-Isopropoxyethyl)piperidine hydrochloride
Supplier: Matrix Scientific
050325-500MG
050325-2.5G
101891-200EA
364.62
USD
101891-200
101891-202
4-(2-Isopropoxyethyl)piperidine hydrochloride
4-(2-Isopropoxyethyl)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₂ClNO MW: 207.75 g/mol |
MDL Number:
MFCD13560726 CAS Number: 1220032-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...