3-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride
Supplier: Matrix Scientific
050324-500MG
050324-2.5G
101891-196EA
364.62
USD
101891-196
101891-198
3-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride
3-{[2-Nitro-4-(trifluoromethyl)phenoxy]methyl}pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₄ClF₃N₂O₃ |
MDL Number:
MFCD13560725 CAS Number: 1220020-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...