2-{2-[2-Nitro-4-(trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050323-500MG 050323-2.5G
101891-192EA 364.62 USD
101891-192 101891-194
2-{2-[2-Nitro-4-(trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
2-{2-[2-Nitro-4-(trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClF₃N₂O₃
MDL Number: MFCD13560724
CAS Number: 1220016-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR