3-{2-[2-Nitro-4-(trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050322-500MG 050322-2.5G
101891-188EA 364.62 USD
101891-188 101891-190
3-{2-[2-Nitro-4-(trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride
3-{2-[2-Nitro-4-(trifluoromethyl)phenoxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈ClF₃N₂O₃
MDL Number: MFCD13560723
CAS Number: 1219967-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR