3-[(2-Chloro-4-nitrophenoxy)methyl]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050320-500MG 050320-2.5G
101891-180EA 364.62 USD
101891-180 101891-182
3-[(2-Chloro-4-nitrophenoxy)methyl]pyrrolidine hydrochloride
3-[(2-Chloro-4-nitrophenoxy)methyl]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄Cl₂N₂O₃
MDL Number: MFCD13560721
CAS Number: 1220017-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR