4-[2-(2-Chloro-4-nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050317-500MG 050317-2.5G
101891-168EA 364.62 USD
101891-168 101891-170
4-[2-(2-Chloro-4-nitrophenoxy)ethyl]piperidine hydrochloride
4-[2-(2-Chloro-4-nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₂N₂O₃
MDL Number: MFCD13560718
CAS Number: 1220032-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR