2-[2-(2-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050315-500MG 050315-2.5G
101891-160EA 186.15 USD
101891-160 101891-162
2-[2-(2-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride
2-[2-(2-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₃N₂O
MDL Number: MFCD13560716
CAS Number: 1219976-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR