2-[2-(3-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050314-500MG 050314-2.5G
101891-156EA 364.62 USD
101891-156 101891-158
2-[2-(3-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride
2-[2-(3-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₃N₂O
MDL Number: MFCD13560715
CAS Number: 1219967-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR