2-[2-(4-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride

Supplier: Matrix Scientific

050313-500MG 050313-2.5G
101891-152EA 364.62 USD
101891-152 101891-154
2-[2-(4-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride
2-[2-(4-Piperidinyl)ethoxy]-5-(trifluoromethyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClF₃N₂O
MDL Number: MFCD13560714
CAS Number: 1220030-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR