2-Bromo-6-(3-pyrrolidinylmethoxy)pyridine hydrochloride

Supplier: Matrix Scientific

050312-500MG 050312-2.5G
101891-148EA 364.62 USD
101891-148 101891-150
2-Bromo-6-(3-pyrrolidinylmethoxy)pyridine hydrochloride
2-Bromo-6-(3-pyrrolidinylmethoxy)pyridine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄BrClN₂O
MW: 293.6 g/mol
MDL Number: MFCD13560713
CAS Number: 1220020-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR