2-Bromo-6-[2-(2-piperidinyl)ethoxy]pyridine hydrochloride

Supplier: Matrix Scientific

050311-500MG 050311-2.5G
101891-144EA 364.62 USD
101891-144 101891-146
2-Bromo-6-[2-(2-piperidinyl)ethoxy]pyridine hydrochloride
2-Bromo-6-[2-(2-piperidinyl)ethoxy]pyridine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈BrClN₂O
MDL Number: MFCD13560712
CAS Number: 1220032-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR