5-Nitro-2-[2-(2-piperidinyl)ethoxy]pyridine hydrochloride
Supplier: Matrix Scientific
050307-500MG
050307-2.5G
101891-128EA
364.62
USD
101891-128
101891-130
5-Nitro-2-[2-(2-piperidinyl)ethoxy]pyridine hydrochloride
5-Nitro-2-[2-(2-piperidinyl)ethoxy]pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O₃ |
MDL Number:
MFCD13560708 CAS Number: 1219976-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...