5-Nitro-2-[2-(3-piperidinyl)ethoxy]pyridine hydrochloride
Supplier: Matrix Scientific
050306-500MG
050306-2.5G
101891-124EA
364.62
USD
101891-124
101891-126
5-Nitro-2-[2-(3-piperidinyl)ethoxy]pyridine hydrochloride
5-Nitro-2-[2-(3-piperidinyl)ethoxy]pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O₃ |
MDL Number:
MFCD13560707 CAS Number: 1219977-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...