5-Nitro-2-[2-(4-piperidinyl)ethoxy]pyridine hydrochloride
Supplier: Matrix Scientific
050305-500MG
050305-2.5G
101891-120EA
364.62
USD
101891-120
101891-122
5-Nitro-2-[2-(4-piperidinyl)ethoxy]pyridine hydrochloride
5-Nitro-2-[2-(4-piperidinyl)ethoxy]pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O₃ |
MDL Number:
MFCD13560706 CAS Number: 1219967-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...