6-Chloro-2-pyrazinyl-2-(2-piperidinyl)ethyl ether hydrochloride

Supplier: Matrix Scientific

050303-500MG 050303-2.5G
101891-112EA 364.62 USD
101891-112 101891-114
6-Chloro-2-pyrazinyl-2-(2-piperidinyl)ethyl ether hydrochloride
6-Chloro-2-pyrazinyl-2-(2-piperidinyl)ethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇Cl₂N₃O
MW: 278.18 g/mol
MDL Number: MFCD13560704
CAS Number: 1220039-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR