6-Chloro-2-pyrazinyl-2-(3-piperidinyl)ethyl ether hydrochloride
Supplier: Matrix Scientific
050302-500MG
050302-2.5G
101891-108EA
364.62
USD
101891-108
101891-110
6-Chloro-2-pyrazinyl-2-(3-piperidinyl)ethyl ether hydrochloride
6-Chloro-2-pyrazinyl-2-(3-piperidinyl)ethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇Cl₂N₃O MW: 278.18 g/mol |
MDL Number:
MFCD13560703 CAS Number: 1220032-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...