6-Chloro-2-pyrazinyl-2-(4-piperidinyl)ethyl ether hydrochloride
Supplier: Matrix Scientific
050301-500MG
050301-2.5G
101891-104EA
364.62
USD
101891-104
101891-106
6-Chloro-2-pyrazinyl-2-(4-piperidinyl)ethyl ether hydrochloride
6-Chloro-2-pyrazinyl-2-(4-piperidinyl)ethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇Cl₂N₃O MW: 278.18 g/mol |
MDL Number:
MFCD13560702 CAS Number: 1219976-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...