6-Chloro-4-pyrimidinyl 2-(2-piperidinyl)ethylether hydrochloride

Supplier: Matrix Scientific

050299-500MG 050299-2.5G
101891-096EA 364.62 USD
101891-096 101891-098
6-Chloro-4-pyrimidinyl 2-(2-piperidinyl)ethylether hydrochloride
6-Chloro-4-pyrimidinyl 2-(2-piperidinyl)ethylether hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇Cl₂N₃O
MDL Number: MFCD13560700
CAS Number: 1219967-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR