6-Chloro-4-pyrimidinyl 2-(4-piperidinyl)ethylether hydrochloride
Supplier: Matrix Scientific
050297-500MG
050297-2.5G
101891-088EA
364.62
USD
101891-088
101891-090
6-Chloro-4-pyrimidinyl 2-(4-piperidinyl)ethylether hydrochloride
6-Chloro-4-pyrimidinyl 2-(4-piperidinyl)ethylether hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇Cl₂N₃O MW: 278.18 g/mol |
MDL Number:
MFCD13560698 CAS Number: 1219977-00-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...