3-{[(4-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050296-500MG 050296-2.5G
101891-084EA 364.62 USD
101891-084 101891-086
3-{[(4-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(4-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560697
CAS Number: 1220038-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR