2-{2-[(4-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050295-500MG 050295-2.5G
101891-080EA 364.62 USD
101891-080 101891-082
2-{2-[(4-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(4-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560696
CAS Number: 1220038-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR