4-{2-[(4-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050293-500MG 050293-2.5G
101891-072EA 364.62 USD
101891-072 101891-074
4-{2-[(4-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(4-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560694
CAS Number: 1220032-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR