3,5-Dimethoxybenzyl-3-pyrrolidinylmethyl ether hydrochloride
Supplier: Matrix Scientific
050292-500MG
050292-2.5G
101891-068EA
364.62
USD
101891-068
101891-070
3,5-Dimethoxybenzyl-3-pyrrolidinylmethyl ether hydrochloride
3,5-Dimethoxybenzyl-3-pyrrolidinylmethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO₃ |
MDL Number:
MFCD13560693 CAS Number: 1219964-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...