2-{2-[(3,5-Dimethoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050291-500MG 050291-2.5G
101891-064EA 364.62 USD
101891-064 101891-066
2-{2-[(3,5-Dimethoxybenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(3,5-Dimethoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO₃
MDL Number: MFCD13560692
CAS Number: 1220038-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR