3-{2-[(3,5-Dimethoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050290-500MG 050290-2.5G
101891-060EA 364.62 USD
101891-060 101891-062
3-{2-[(3,5-Dimethoxybenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3,5-Dimethoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO₃
MDL Number: MFCD13560691
CAS Number: 1220038-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR