3-{[(3,5-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050288-500MG 050288-2.5G
101891-052EA 364.62 USD
101891-052 101891-054
3-{[(3,5-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(3,5-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride
Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13560689
CAS Number: 1219976-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR