3-{2-[(3,5-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050286-500MG 050286-2.5G
101891-044EA 364.62 USD
101891-044 101891-046
3-{2-[(3,5-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3,5-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClF₂NO
MDL Number: MFCD13560687
CAS Number: 1219976-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR