4-{2-[(3,5-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050285-500MG 050285-2.5G
101891-040EA 364.62 USD
101891-040 101891-042
4-{2-[(3,5-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3,5-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClF₂NO
MDL Number: MFCD13560686
CAS Number: 1220038-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR