3-{[(3-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050284-500MG 050284-2.5G
101891-036EA 364.62 USD
101891-036 101891-038
3-{[(3-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(3-Methoxybenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD13560685
CAS Number: 1220032-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR