3-{2-[(3-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050282-500MG 050282-2.5G
101891-028EA 364.62 USD
101891-028 101891-030
3-{2-[(3-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride
3-{2-[(3-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560683
CAS Number: 1220016-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR