4-{2-[(3-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050281-500MG 050281-2.5G
101891-024EA 364.62 USD
101891-024 101891-026
4-{2-[(3-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride
4-{2-[(3-Methoxybenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO₂
MDL Number: MFCD13560682
CAS Number: 1220038-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR