3-{[(2,6-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Supplier: Matrix Scientific

050280-500MG 050280-2.5G
101891-020EA 364.62 USD
101891-020 101891-022
3-{[(2,6-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride
3-{[(2,6-Difluorobenzyl)oxy]methyl}pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClF₂NO
MDL Number: MFCD13560681
CAS Number: 1220016-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR