2-{2-[(2,6-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Supplier: Matrix Scientific

050279-500MG 050279-2.5G
101891-016EA 364.62 USD
101891-016 101891-018
2-{2-[(2,6-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride
2-{2-[(2,6-Difluorobenzyl)oxy]ethyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClF₂NO
MDL Number: MFCD13560680
CAS Number: 1220032-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR